MilliporeSigma
2,6-Dimethylphenol: a new metabolite of mexiletine.
Research communications in chemical pathology and pharmacology (1987-10-01)
O Grech-Bélanger, J Turgeon, M Lalande
ABSTRACT
A new metabolite, 2,6-dimethylphenol obtained by O-dealkylation of mexiletine by rabbit liver was identified. A g.c.-f.i.d. method was developed for its analysis in hepatic homogenates and some of the characteristics of the metabolic reaction were studied. Formation of 2,6-dimethylphenol is microsomal and is optimal under aerobic conditions. Both carbon monoxide and n-octylamine significantly depress this O-dealkylation reaction.
Sigma-Aldrich
2,6-Xylenol, ≥99%, FG
Sigma-Aldrich
2,6-Dimethylphenol, 99%
Supelco
2,6-Dimethylphenol, PESTANAL®, analytical standard
Sigma-Aldrich
2,6-Dimethylphenol, ≥99.5%