MilliporeSigma
  • Identification of N-beta-L-aspartyl-L-phenylalanine as a normal constituent of human plasma and urine.
Identification of N-beta-L-aspartyl-L-phenylalanine as a normal constituent of human plasma and urine.
The Journal of nutrition (1989-05-01)
E G Burton, G L Schoenhard, J A Hill, R E Schmidt, J D Hribar, F N Kotsonis, J A Oppermann
ABSTRACT
A new beta-aspartyl dipeptide, N-beta-L-aspartyl-L-phenylalanine (beta-AP), has been isolated and identified in urine and plasma from normal human volunteers. beta-AP was isolated from urine samples by high performance liquid chromatography (HPLC). Its identity and stereochemistry were demonstrated by HPLC and gas chromatography/mass spectrometry (GC-MS). The mean urinary beta-AP concentration in the subjects was 0.63 +/- 0.14 microgram/mg creatinine when averaged over two consecutive days of urine collection. Daily beta-AP excretion, determined from two 24-h urine samples collected from five individuals, was 801 +/- 117 micrograms/d (2.7 mumol/d). No diurnal rhythm was evident within the 24-h collection periods. beta-AP was also isolated from human plasma by HPLC and identified by GC-MS. Plasma from subjects contained approximately 5 ng beta-AP/ml. Furthermore, beta-AP was formed when asparagine and phenylalanine were incubated with an enzyme extract from human kidney. Thus, at least some of the beta-AP present in humans, and presumably other beta-aspartyl dipeptides as well, appears to be synthesized endogenously.
Sigma-Aldrich
Asp-Phe, 96%