MilliporeSigma
  • Medical management of endometriosis: emerging evidence linking inflammation to disease pathophysiology.
Medical management of endometriosis: emerging evidence linking inflammation to disease pathophysiology.
Minerva ginecologica (2013-04-20)
K L Bruner-Tran, J L Herington, A J Duleba, H S Taylor, K G Osteen
ABSTRACT
Progesterone action normally mediates the balance between anti-inflammatory and proinflammatory processes throughout the female reproductive tract. However, in women with endometriosis, endometrial progesterone resistance, characterized by alterations in progesterone responsive gene and protein expression, is now considered a central element in disease pathophysiology. Recent studies additionally suggest that the peritoneal microenvironment of endometriosis patients exhibits altered physiological characteristics that may further promote inflammation-driven disease development and progression. Within this review, we summarize our current understanding of the pathogenesis of endometriosis with an emphasis on the role that inflammation plays in generating not only the progesterone-resistant eutopic endometrium but also a peritoneal microenvironment that may contribute significantly to disease establishment. Viewing endometriosis from the emerging perspective that a progesterone resistant endometrium and an immunologically compromised peritoneal microenvironment are biologically linked risk factors for disease development provides a novel mechanistic framework to identify new therapeutic targets for appropriate medical management.
Product Description
Supelco
Progesterone, VETRANAL®, analytical standard
Sigma-Aldrich
Progesterone, ≥99%
Sigma-Aldrich
Progesterone, γ-irradiated, BioXtra, suitable for cell culture
Sigma-Aldrich
Progesterone, powder, BioReagent, suitable for cell culture
Supelco
Progesterone, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Progesterone, meets USP testing specifications
Supelco
Progesterone solution, 1.0 mg/mL in acetonitrile, ampule of 1 mL, certified reference material, Cerilliant®