MilliporeSigma
Cytoskeleton: axons earn their stripes.
Current biology : CB (2013-03-12)
Matthew N Rasband
ABSTRACT
Axons must be supported by a strong and flexible cytoskeleton. New 'super-resolution' imaging of the submembranous axonal cytoskeleton reveals that it is organized in a periodic, ladder-like structure with alternating rings of actin linked together by intervening complexes of spectrin.
Sigma-Aldrich
Spectrin from human erythrocytes, buffered aqueous glycerol solution