MilliporeSigma
  • Toward high-energy-density, high-efficiency, and moderate-temperature chip-scale thermophotovoltaics.
Toward high-energy-density, high-efficiency, and moderate-temperature chip-scale thermophotovoltaics.
Proceedings of the National Academy of Sciences of the United States of America (2013-02-27)
Walker R Chan, Peter Bermel, Robert C N Pilawa-Podgurski, Christopher H Marton, Klavs F Jensen, Jay J Senkevich, John D Joannopoulos, Marin Soljacic, Ivan Celanovic
ABSTRACT
The challenging problem of ultra-high-energy-density, high-efficiency, and small-scale portable power generation is addressed here using a distinctive thermophotovoltaic energy conversion mechanism and chip-based system design, which we name the microthermophotovoltaic (μTPV) generator. The approach is predicted to be capable of up to 32% efficient heat-to-electricity conversion within a millimeter-scale form factor. Although considerable technological barriers need to be overcome to reach full performance, we have performed a robust experimental demonstration that validates the theoretical framework and the key system components. Even with a much-simplified μTPV system design with theoretical efficiency prediction of 2.7%, we experimentally demonstrate 2.5% efficiency. The μTPV experimental system that was built and tested comprises a silicon propane microcombustor, an integrated high-temperature photonic crystal selective thermal emitter, four 0.55-eV GaInAsSb thermophotovoltaic diodes, and an ultra-high-efficiency maximum power-point tracking power electronics converter. The system was demonstrated to operate up to 800 °C (silicon microcombustor temperature) with an input thermal power of 13.7 W, generating 344 mW of electric power over a 1-cm(2) area.
Sigma-Aldrich
Propane, 99.5%
Sigma-Aldrich
Propane, 98%