MilliporeSigma
  • Mussel-inspired green synthesis of silver nanoparticles on graphene oxide nanosheets for enhanced catalytic applications.
Mussel-inspired green synthesis of silver nanoparticles on graphene oxide nanosheets for enhanced catalytic applications.
Chemical communications (Cambridge, England) (2013-02-26)
Eun Kyung Jeon, Eunyong Seo, Eunhee Lee, Wonoh Lee, Moon-Kwang Um, Byeong-Su Kim
ABSTRACT
We report a facile green approach to the synthesis of silver nanoparticles (Ag NPs) on the surface of graphene oxide nanosheets functionalized with mussel-inspired dopamine (GO-Dopa) without additional reductants or stabilizers at room temperature. The resulting hybrid Ag/GO-Dopa exhibits good dispersity and excellent catalytic activity in the reduction of nitroarenes.
Sigma-Aldrich
3,4-Dihydroxy-DL-phenylalanine