MilliporeSigma
  • [Content determination of two isomers containd in Garcinia hanburyi by HPLC].
[Content determination of two isomers containd in Garcinia hanburyi by HPLC].
Zhongguo Zhong yao za zhi = Zhongguo zhongyao zazhi = China journal of Chinese materia medica (2013-02-13)
Jun-Yan Zhang, Guang-Ping Xia, Na-Xia Zhao, Yun-Ping Chang, Ying-Mei Han
ABSTRACT
To establish a method for determing the content of two isomers containd in Garcinia hanburyi by HPLC. Chromatographic column of SunFire (Waters) C8 (2.1 mm x 150 mm, 3.5 microm) was adopted, with acetonitrile-methanol-0.3% trifluoroacetic acid (36: 37:27) as the mobile phase. The detection wavelength was 360 nm,the flow rate was 0.3 mL x min(-1), and the column temperature was 28 degrees C. The linear regression equation of r-gambogic acid was Y = 2.87 x 10(6) X - 2.24 x 10(5), r = 0.999 9. The linear regression equation of S-gambogic acid was Y = 3.31 x 10(6) X - 1.44 x 10(5), r = 0.999 9. The average recoveries were 100.0% and 100.9%, with RSD being 2.1% and 2.5% (n = 6), respectivley. The average contents of two gambogic acid in G. hanburyi were 30.06% and 21.45%, respectively. The method was so convenient and stable that it can be used for identification and content determination of two isomers containd in G. hanburyi.
Product Description
Supelco
Discovery® C8 HPLC Column, 5 μm particle size, L × I.D. 25 cm × 4.6 mm
Supelco
Discovery® C8 HPLC Column, 5 μm particle size, L × I.D. 25 cm × 3 mm
Supelco
Discovery® C8 HPLC Column, 5 μm particle size, L × I.D. 5 cm × 2.1 mm
Supelco
Discovery® C8 HPLC Column, 5 μm particle size, L × I.D. 15 cm × 2.1 mm
Supelco
Discovery® C8 Supelguard Guard Cartridge, 5 μm particle size, L × I.D. 2 cm × 2.1 mm
Supelco
Discovery® C8 Supelguard Guard Cartridge, 5 μm particle size, L × I.D. 2 cm × 4 mm
Supelco
Discovery® C8 HPLC Column, 5 μm particle size, L × I.D. 5 cm × 4.6 mm
Supelco
Discovery® C8 HPLC Column, 5 μm particle size, L × I.D. 12.5 cm × 4 mm
Supelco
Discovery® C8 Supelguard Guard Cartridge, 5 μm particle size, L × I.D. 2 cm × 4 mm
Supelco
Discovery® C8 HPLC Column, 5 μm particle size, L × I.D. 12.5 cm × 4.6 mm
Supelco
Discovery® C8 HPLC Column, 5 μm particle size, L × I.D. 15 cm × 4.6 mm