MilliporeSigma
Direct bandgap narrowing in Ge LED's on Si substrates.
Optics express (2013-02-08)
Michael Oehme, Martin Gollhofer, Daniel Widmann, Marc Schmid, Mathias Kaschel, Erich Kasper, Jörg Schulze
ABSTRACT
In this paper we investigate the influence of n-type doping in Ge light emitting diodes on Si substrates on the room temperature emission spectrum. The layer structures are grown with a special low temperature molecular beam epitaxy process resulting in a slight tensile strain of 0.13%. The Ge LED's show a dominant direct bandgap emission with shrinking bandgap at the Γ point in dependence of n-type doping level. The emission shift (38 meV at 10²⁰cm⁻³) is mainly assigned to bandgap narrowing at high doping. The electroluminescence intensity increases with doping concentrations up to 3x10¹⁹cm⁻³ and decreases sharply at higher doping levels. The integrated direct gap emission intensity increases superlinear with electrical current density. Power exponents vary from about 2 at low doping densities up to 3.6 at 10²⁰cm⁻³ doping density.
Sigma-Aldrich
Germanium, chips, 99.999% trace metals basis
Sigma-Aldrich
Germanium, powder, −100 mesh, ≥99.999% trace metals basis
Sigma-Aldrich
Germanium, chips, 99.999% trace metals basis
Sigma-Aldrich
Germanium, powder, −100 mesh, ≥99.99% trace metals basis