MilliporeSigma
  • Fabrication of porous polyvinyl alcohol scaffold for bone tissue engineering via selective laser sintering.
Fabrication of porous polyvinyl alcohol scaffold for bone tissue engineering via selective laser sintering.
Biofabrication (2013-02-07)
Cijun Shuai, Zhongzheng Mao, Haibo Lu, Yi Nie, Huanlong Hu, Shuping Peng
ABSTRACT
A tetragonal polyvinyl alcohol (PVA) scaffold with 3D orthogonal periodic porous architecture was fabricated via selective laser sintering (SLS) technology. The scaffold was fabricated under the laser power of 8 W, scan speed of 600 mm min(-1), laser spot diameter of 0.8 mm and layer thickness of 0.15 mm. The microstructure analysis showed that the degree of crystallization decreased while the PVA powder melts gradually and fuses together completely with laser power increasing. Thermal decomposition would occur if the laser power was further higher (16 W or higher in the case). The porous architecture was controllable and totally interconnected. The porosity of the fabricated scaffolds was measured to be 67.9 ± 2.7% which satisfies the requirement of micro-pores of the bone scaffolds. Its bioactivity and biocompatibility were also evaluated in vitro as tissue engineering (TE) scaffolds. In vitro adhesion assay showed that the amount of pores increased while the scaffold remains stable and intact after immersion in simulated body fluid for seven days. Moreover, the number of MG-63 cells and the bridge between cells increased with increasing time in cell culture. The present work demonstrates that PVA scaffolds with well-defined porous architectures via SLS technology were designed and fabricated for bone TE.
Sigma-Aldrich
Mowiol® 10-98, Mw ~61,000
Sigma-Aldrich
Poly(vinyl alcohol), average Mw 13,000-23,000, 98% hydrolyzed
Sigma-Aldrich
Mowiol® 4-98, Mw ~27,000
Sigma-Aldrich
Mowiol® 8-88, Mw ~67,000
Sigma-Aldrich
Poly(vinyl alcohol), Mw 85,000-124,000, 99+% hydrolyzed
Sigma-Aldrich
Poly(vinyl alcohol), average Mw 146,000-186,000, 87-89% hydrolyzed
Sigma-Aldrich
Poly(vinyl alcohol), Mw 31,000-50,000, 98-99% hydrolyzed
Sigma-Aldrich
Poly(vinyl alcohol), Mw 9,000-10,000, 80% hydrolyzed
Sigma-Aldrich
Poly(vinyl alcohol), average Mw 31,000-50,000, 87-89% hydrolyzed
Sigma-Aldrich
Poly(vinyl alcohol), Mw 146,000-186,000, 99+% hydrolyzed
Sigma-Aldrich
Poly(vinyl alcohol), average Mw 85,000-124,000, 87-89% hydrolyzed
Sigma-Aldrich
Poly(vinyl alcohol), Mw 89,000-98,000, 99+% hydrolyzed
Sigma-Aldrich
Poly(vinyl alcohol), Mw 13,000-23,000, 87-89% hydrolyzed
Sigma-Aldrich
Poly(vinyl alcohol), 87-90% hydrolyzed, average mol wt 30,000-70,000
Sigma-Aldrich
Poly(vinyl alcohol), Fully hydrolyzed
Sigma-Aldrich
Poly(vinyl alcohol), average Mw 130,000, 99+% hydrolyzed
Sigma-Aldrich
Poly(vinyl alcohol), SAJ first grade, degree of polymerization 500
Sigma-Aldrich
Mowiol® 6-98, Mw ~47,000
Sigma-Aldrich
Mowiol® 20-98, Mw ~125,000
Sigma-Aldrich
Mowiol® 4-88, Mw ~31,000
Sigma-Aldrich
Mowiol® 28-99, Mw ~145,000
Sigma-Aldrich
Mowiol® 18-88, Mw ~130,000
Sigma-Aldrich
Mowiol® 40-88, average Mw ~205,000 g/mol
Sigma-Aldrich
Kuraray Poval ®56-98, Mw ~195,000