MilliporeSigma
  • Self-assembly of mesoporous CuO nanosheets-CNT 3D-network composites for lithium-ion batteries.
Self-assembly of mesoporous CuO nanosheets-CNT 3D-network composites for lithium-ion batteries.
Nanoscale (2013-01-31)
Hongwen Huang, Yu Liu, Junhua Wang, Mingxia Gao, Xinsheng Peng, Zhizhen Ye
ABSTRACT
A facile, flexible and large-scale technique was proposed to prepare a CuO-CNT 3D-network composite with the aid of electrostatic interactions in aqueous solution. The composite greatly improves the electrochemical performance. At a rate of 0.1 C, the cycling discharge capacity of the optimal composite is more than 2.3 times of that of unmodified mesoporous CuO nanosheets as the active material in an anode after 40 cycles.
Sigma-Aldrich
Copper(II) oxide, powder, 99.995% trace metals basis
Sigma-Aldrich
Copper(II) oxide, powder, 99.99% trace metals basis
Sigma-Aldrich
Copper(II) oxide, nanopowder, <50 nm particle size (TEM)
Sigma-Aldrich
Copper(II) oxide, 99.999% trace metals basis
Sigma-Aldrich
Copper(II) oxide, ACS reagent, ≥99.0%
Sigma-Aldrich
Copper(II) oxide, needles, mixture of CuO and Cu2O, ACS reagent
Sigma-Aldrich
Copper(II) oxide, powder, <10 μm, 98%
Sigma-Aldrich
Copper(II) oxide on alumina, 14-30 mesh, extent of labeling: 13 wt. % loading