MilliporeSigma
  • Colloidal aggregates of Pd nanoparticles supported by larch arabinogalactan.
Colloidal aggregates of Pd nanoparticles supported by larch arabinogalactan.
The journal of physical chemistry. B (2013-01-31)
Ekaterina R Gasilova, Galina N Matveeva, Galina P Aleksandrova, Boris G Sukhov, Boris A Trofimov
ABSTRACT
Palladium nanoparticles (PdNPs) are used in catalysis, hydrogen storage, biomedicine, and so on. Arranging the self-assembly of PdNPs within colloidal aggregates is desirable for improving their consumer properties. Stable widely dispersed colloidal aggregates of larch arabinogalactan (LARB) containing nanosized (5-nm) PdNPs were obtained by reducing Pd ions in alkaline solutions of LARB. Centrifugation resulted in a set of LARB-PdNP colloids ranging from 60 to 240 nm. The colloids were studied by static light scattering (SLS) and dynamic light scattering (DLS). The SLS data presented as Kratki plots correspond to a particle scattering factor of linear rather than branched chains. The fractal dimension of the LARB-PdNP colloids was found by SLS to be d = 1.96, which is between the values for diffusion- and reaction-limited aggregation. This result is ascribed to the aggregate's internal motion, which is evident from the power-law exponent of the dependence of the DLS relaxation rate on the scattering vector, <Γ> ~ q(α) with α = 2.24. The structure-sensitive ratio of the radius of gyration to the hydrodynamic radius was found to vary within the interval of 0.8 ≤ R(g)/R(h) ≤ 1.2 corresponding, to the spherical form of LARB-PdNP colloids. A spiderweblike PdNP distribution pattern was observed by transmission electron microscopy. Insertion of PdNPs did not affect the fractal dimension, the power-law exponent α, or the architecture of the pristine LARB aggregates in water. The red shift of the surface plasmon extinction observed with increasing LARB-PdNP colloidal size indicates the collective optical response of the PdNP ensemble in the colloid.
Sigma-Aldrich
(+)-Arabinogalactan, ≥80% (HPLC), from larch wood
Supelco
(+)-Arabinogalactan, for use with Total Dietary Fiber Control Kit, TDF-C10, from larch wood