MilliporeSigma
  • Synthesis of 2-hydroxy-1,4-oxazin-3-ones through ring transformation of 3-hydroxy-4-(1,2-dihydroxyethyl)-β-lactams and a study of their reactivity.
Synthesis of 2-hydroxy-1,4-oxazin-3-ones through ring transformation of 3-hydroxy-4-(1,2-dihydroxyethyl)-β-lactams and a study of their reactivity.
Chemistry (Weinheim an der Bergstrasse, Germany) (2013-01-26)
Karen Mollet, Hannelore Goossens, Nicola Piens, Saron Catak, Michel Waroquier, Karl W Törnroos, Veronique Van Speybroeck, Matthias D'hooghe, Norbert De Kimpe
ABSTRACT
The reactivity of 3-hydroxy-4-(1,2-dihydroxyethyl)-β-lactams with regard to the oxidant sodium periodate was evaluated, unexpectedly resulting in the exclusive formation of new 2-hydroxy-1,4-oxazin-3-ones through a C3C4 bond cleavage of the intermediate 4-formyl-3-hydroxy-β-lactams followed by a ring expansion. This peculiar transformation stands in sharp contrast with the known NaIO(4)-mediated oxidation of 3-alkoxy- and 3-phenoxy-4-(1,2-dihydroxyethyl)-β-lactams, which exclusively leads to the corresponding 4-formyl-β-lactams without a subsequent ring enlargement. In addition, this new class of functionalized oxazin-3-ones was further evaluated for its potential use as building blocks in the synthesis of a variety of differently substituted oxazin-3-ones, morpholin-3-ones and pyrazinones. Furthermore, additional insights into the mechanism and the factors governing this new ring-expansion reaction were provided by means of density functional theory calculations.
Sigma-Aldrich
Sodium (meta)periodate, BioUltra, ≥99.5% (RT)
Sigma-Aldrich
Sodium (meta)periodate, ≥99.0%
Sigma-Aldrich
Periodic acid, SAJ special grade, ≥98.5%
Sigma-Aldrich
Periodic acid, 99.999% trace metals basis
Sigma-Aldrich
Periodic acid, puriss. p.a., ACS reagent, crystallized, ≥99.0% (RT)
Sigma-Aldrich
Sodium periodate, ACS reagent, ≥99.8%
Sigma-Aldrich
Periodic acid, ACS reagent, 99%
Sigma-Aldrich
Periodic acid, suitable for electrophoresis, ≥99%
Sigma-Aldrich
Periodic acid, ReagentPlus®, ≥99.0%