MilliporeSigma
  • Strategies for endogenous spinal cord repair: HPMA hydrogel to recruit migrating endogenous stem cells.
Strategies for endogenous spinal cord repair: HPMA hydrogel to recruit migrating endogenous stem cells.
Advances in experimental medicine and biology (2013-01-03)
Araceli Espinosa-Jeffrey, Karlos Oregel, Laurent Wiggins, Remelyn Valera, Kathrin Bosnoyan, Chioma Agbo, Oluwole Awosika, Paul M Zhao, Jean de Vellis, Stéphane Woerly
ABSTRACT
Injury to the spinal cord disrupts ascending and descending axonal pathways and causes tissue damage with a subsequent limited cellular regeneration. Successful treatment would encompass the restoration of the cytoarchitecture, homeostasis and function all in dear need. Transplantation-based treatments using exogenous cells are the most favoured approach. Yet, with the advent of the stem cell concept and continuous progress in the field it became clear that the endogenous potential for repair is greater than previously thought. As an alternative to neural grafting, we and other researchers have aimed at understanding what are the elements needed for a successful repair with self progenitors that would give rise to the cell types needed to restore function of the central nervous system. Some studies involve both scaffolds and cell grafts. Here we describe studies on spinal cord repair using what we call "endogenous tissue engineering for regenerative medicine". The approach involves a hydrogel that mimics the natural milieu where endogenous pre-existing and newly formed cells populate the gel progressively allowing for the integration of CNS self populations leading to a successful recovery of function. Highlight aspects learned from this type of studies are that: Endogenous reconstruction of the injured spinal cord is possible by using the adequate support. The contribution of nestin-expressing progenitors to spinal cord regeneration is continuous and substantial both, in the reconstructed segment as well as, along the distal and caudal segments of the reconstructed spinal cord. Most of these cells appear to have been in a quiescent state until the injury occurred and only a small fraction of these neural progenitors was produced via cell proliferation. The hydrogel combined with exercise was necessary and sufficient to restore locomotor function in cats that underwent spinal transaction followed by reconstructive surgery. This recovery of function was first seen 28 days after surgery and continued to improve for at least 21 months. Therefore, endogenous pre-existing and newly formed cells populated the gel scaffold established contact with the non injured tissue and lead to recovery of function.
Sigma-Aldrich
Hydroxypropyl methacrylate, Mixture of hydroxypropyl and hydroxyisopropyl methacrylates, 97%, contains 180-220 ppm monomethyl ether hydroquinone as inhibitor