MilliporeSigma
  • Photocatalytic degradation of methyl orange dye using silver (Ag) nanoparticles synthesized from Ulva lactuca.
Photocatalytic degradation of methyl orange dye using silver (Ag) nanoparticles synthesized from Ulva lactuca.
Colloids and surfaces. B, Biointerfaces (2012-12-26)
P Kumar, M Govindaraju, S Senthamilselvi, K Premkumar
ABSTRACT
In this paper, we report on biosynthesis of silver nanoparticles using Ulva lactuca (seaweed) at room temperature along with photocatalytic degradation of methyl orange dye. UV spectral analysis showed peak at 430 nm with special reference to the excitation of surfaces plasmon vibration by silver nanoparticles. FT-IR studies reveal the presence of bioactive functional groups such as phenolic compounds, amines and aromatic ring are found to be the capping and stabilizing agents of nanoparticles. The morphology of silver nanoparticles was found to be spherical and ranges about 48.59 nm as confirmed by HR-SEM. Negative zeta potential value of -34 mV suggests that the nanoparticles are highly stable in colloidal solution. XRD patterns also suggest the occurrence of spherical shaped particles due to the presence of silver ions. Further, photocatalytic degradation of methyl orange was measured spectrophotometrically by using silver as nanocatalyst under visible light illumination. The results revealed that biosynthesized silver nanoparticles using U. lactuca was found to be impressive in degrading methyl orange.
Sigma-Aldrich
Methyl Orange solution, pH 3.1-4.4
Sigma-Aldrich
Methyl Orange solution, pH 3.1-4.4
Sigma-Aldrich
Methyl Orange, for microscopy (Hist.), indicator (pH 3.0-4.4)
Sigma-Aldrich
Methyl Orange, ACS reagent, Dye content 85 %
Sigma-Aldrich
Methyl Orange, SAJ first grade
Sigma-Aldrich
Methyl Orange, JIS special grade