MilliporeSigma
  • One-pot synthesis of (3-sulfopropyl methacrylate potassium)-silica hybrid monolith via thiol-ene click chemistry for CEC.
One-pot synthesis of (3-sulfopropyl methacrylate potassium)-silica hybrid monolith via thiol-ene click chemistry for CEC.
Electrophoresis (2012-11-23)
Huihui Yang, Yingzhuang Chen, Yixuan Liu, Lihua Nie, Shouzhuo Yao
ABSTRACT
A novel (3-sulfopropyl methacrylate potassium)-silica hybrid monolithic column for CEC has been prepared by a simple one-pot approach based on efficient thiol-ene click chemistry. In this process, the polycondensation of hydrolyzed alkoxysilanes and in situ click reaction of vinyl groups on 3-sulfopropyl methacrylate potassium and thiol groups on the precondensed siloxanes simultaneously occurred in a pretreated capillary. Homogeneous monolithic matrix with large through-pores tightly bonded to the inner wall of the capillary was shown by optical microscope and SEM. The minimum plate height of this hybrid monolithic column was determined as 3.9 μm for thiourea. Anilines, alkylbenzenes, and phenols were well separated on this hybrid monolithic column by CEC, which indicated typical reversed-phase and cation-exchange chromatographic retention mechanisms of the column.
Sigma-Aldrich
Aniline, ACS reagent, ≥99.5%
Sigma-Aldrich
Aniline-4-13C, 99 atom % 13C
Sigma-Aldrich
Aniline, ReagentPlus®, 99%
Sigma-Aldrich
Aniline-15N, 98 atom % 15N
Supelco
Aniline, analytical standard
Sigma-Aldrich
Aniline, JIS special grade, ≥99.0%
Sigma-Aldrich
Aniline, SAJ first grade, ≥99.0%
Supelco
Aniline solution, certified reference material, 5000 μg/mL in methanol
Sigma-Aldrich
Aniline-1-13C, 99 atom % 13C
Sigma-Aldrich
Aniline hydrochloride, ≥99%