MilliporeSigma
  • Electrospun eggroll-like CaSnO3 nanotubes with high lithium storage performance.
Electrospun eggroll-like CaSnO3 nanotubes with high lithium storage performance.
Nanoscale (2012-11-14)
Linlin Li, Shengjie Peng, Yan Ling Cheah, Jin Wang, Peifen Teh, Yahwen Ko, Chuiling Wong, Madhavi Srinivasan
ABSTRACT
Novel eggroll-like CaSnO(3) nanotubes have been prepared by a single spinneret electrospinning method followed by calcination in air for the first time. The electrospun sample as a lithium-ion battery electrode material exhibited improved cycling stability and rate capability by virtue of the high surface area and unique hollow interior structure, compared to nanorod-structured CaSnO(3).
Sigma-Aldrich
Tin(IV) oxide, nanopowder, ≤100 nm avg. part. size
Sigma-Aldrich
Tin(IV) oxide, ≥99.99% trace metals basis
Sigma-Aldrich
Tin(IV) oxide, −325 mesh, 99.9% trace metals basis