MilliporeSigma
  • A novel sensing technique for measurement of magnitude and polarity of electrostatic charge distribution across individual particles.
A novel sensing technique for measurement of magnitude and polarity of electrostatic charge distribution across individual particles.
International journal of pharmaceutics (2012-10-16)
Tariq Hussain, Waseem Kaialy, Tong Deng, Mike S A Bradley, Ali Nokhodchi, David Armour-Chélu
ABSTRACT
Electrostatic charge is generated during powder handling due to particle-particle and particle-wall collisions, rubbing, sliding, and rolling. In case of bipolar charge generation, the electrostatic forces may significantly change the inner forces and increase powder adhesion and cause a serious problem in material handling process. Therefore, the knowledge of distribution of charge across the individual particles is helpful to identify the role of triboelectrification and the effects of various relevant variables especially change in the contact materials, environmental conditions during processing, etc. A novel approach based on inductive sensor has been developed to detect the either polarity of charged particle and to characterise the bipolar charge distribution in the population of particulate material. To achieve this, an amplification unit configured as a pure integrator and signal processing techniques has been used to de-noise and correct the baseline of signal and MATLAB algorithm developed for peak detection. The polarity of charged particles obtained by this method is calibrated with Faraday pail method and the results are promising. Experimental study has been carried out by using two distinct populations of oppositely charged particles (glass beads-PVC, olivine sand, and silica sand). The obtained results indicate that the method is able to detect the distribution of polarities of charged particles.
Sigma-Aldrich
Poly(vinyl chloride), average Mw ~43,000, average Mn ~22,000
Sigma-Aldrich
Poly(vinyl chloride), average Mw ~233,000, average Mn ~99,000
Sigma-Aldrich
Poly(vinyl chloride), average Mw ~62,000, average Mn ~35,000
Poly(vinyl chloride), European Pharmacopoeia (EP) Reference Standard
Supelco
Poly(vinyl chloride), Selectophore, high molecular weight
Sigma-Aldrich
Poly(vinyl chloride), low molecular weight
Supelco
Poly(vinyl chloride), analytical standard, average Mw 85,000 (Typical), average Mn 40,000 (Typical)
Sigma-Aldrich
Poly(vinyl chloride), high molecular weight