MilliporeSigma
  • In situ preparation of novel p-n junction photocatalyst BiOI/(BiO)2CO3 with enhanced visible light photocatalytic activity.
In situ preparation of novel p-n junction photocatalyst BiOI/(BiO)2CO3 with enhanced visible light photocatalytic activity.
Journal of hazardous materials (2012-09-29)
Jing Cao, Xin Li, Haili Lin, Shifu Chen, Xianliang Fu
ABSTRACT
Novel p-n junction photocatalysts BiOI/(BiO)2CO3 with different contents of BiOI were in situ synthesized by etching (BiO)2CO3 precursor with hydroiodic acid (HI) solution. X-ray diffraction (XRD), field emission scanning electron microscopy (FE-SEM), Fourier transform infrared spectrometry (FT-IR), energy-dispersive spectroscopy (EDS) and UV-vis diffuse reflectance spectroscopy (DRS) were employed to study the structures, morphologies and optical properties of the as-prepared samples. Under visible light (λ>420 nm), BiOI/(BiO)2CO3 hybrid displayed much higher photocatalytic activity than pure (BiO)2CO3 and BiOI for the degradation of methyl orange (MO). The increased photocatalytic activity of BiOI/(BiO)2CO3 could be attributed to the formation of the p-n junction between p-BiOI and n-(BiO)2CO3, which effectively suppresses the recombination of photoinduced electron-hole pairs. Moreover, the tests of radical scavengers confirmed that •O2- and h+ were the main reactive species for the degradation of MO.
Sigma-Aldrich
Methyl Orange, JIS special grade
Sigma-Aldrich
Methyl Orange, SAJ first grade
Sigma-Aldrich
Methyl Orange, for microscopy (Hist.), indicator (pH 3.0-4.4)
Sigma-Aldrich
Methyl Orange, ACS reagent, Dye content 85 %
Sigma-Aldrich
Methyl Orange solution, pH 3.1-4.4
Sigma-Aldrich
Methyl Orange solution, pH 3.1-4.4