MilliporeSigma
  • Magnetically tunable elasticity for magnetic hydrogels consisting of carrageenan and carbonyl iron particles.
Magnetically tunable elasticity for magnetic hydrogels consisting of carrageenan and carbonyl iron particles.
The journal of physical chemistry. B (2012-09-15)
Tetsu Mitsumata, Atomu Honda, Hiroki Kanazawa, Mika Kawai
ABSTRACT
A new class of magnetoelastic gel that demonstrates drastic and reversible changes in storage modulus without using strong magnetic fields was obtained. The magnetic gel consists of carrageenan and carbonyl iron particles. The magnetic gel with a volume fraction of magnetic particles of 0.30 exhibited a reversible increase by a factor of 1400 of the storage modulus upon a magnetic field of 500 mT, which is the highest value in the past for magnetorheological soft materials. It is considered that the giant magnetoelastic behavior is caused by both high dispersibility and high mobility of magnetic particles in the carrageenan gel. The off-field storage modulus of the magnetic gel at volume fractions below 0.30 obeyed the Krieger-Dougherty equation, indicating random dispersion of magnetic particles. At 500 mT, the storage modulus was higher than 4.0 MPa, which is equal to that of magnetic fluids, indicating that the magnetic particles move and form a chain structure by magnetic fields. Morphological study revealed the evidence that the magnetic particles embedded in the gel were aligned in the direction of magnetic fields, accompanied by stretching of the gel network. We conclude that the giant magnetoelastic phenomenon originates from the chain structure consisting of magnetic particles similar to magnetic fluids.
Sigma-Aldrich
Iron(0) pentacarbonyl
Sigma-Aldrich
Iron(0) pentacarbonyl, >99.99% trace metals basis