MilliporeSigma
  • Fabrication of polypyrrole/graphene oxide composite nanosheets and their applications for Cr(VI) removal in aqueous solution.
Fabrication of polypyrrole/graphene oxide composite nanosheets and their applications for Cr(VI) removal in aqueous solution.
PloS one (2012-08-29)
Shangkun Li, Xiaofeng Lu, Yanpeng Xue, Junyu Lei, Tian Zheng, Ce Wang
ABSTRACT
In this paper, we report on the simple, reliable synthesis of polypyrrole (PPy)/graphene oxide (GO) composite nanosheets by using sacrificial-template polymerization method. Herein, MnO(2) nanoslices were chosen as a sacrificial-template to deposit PPy, which served as the oxidant as well. During the polymerization of pyrrole on surface of GO nanosheets, MnO(2) component was consumed incessantly. As a result, the PPy growing on the surface of GO nanosheets has the morphology just like the MnO(2) nanoslices. This method can provide the fabrication of PPy nanostructures more easily than conventional route due to its independence of removing template, which usually is a complex and tedious experimental process. The as-prepared PPy/GO composite nanosheets exhibited an enhanced properties for Cr(VI) ions removal in aqueous solution based on the synergy effect. The adsorption capacity of the PPy/GO composite nanosheets is about two times as large as that of conventional PPy nanoparticles. We believe that our findings can open a new and effective avenue to improve the adsorption performance in removing heavy metal ions from waste water.
Sigma-Aldrich
Magnesium oxide, nanopowder, ≤50 nm particle size (BET)
Sigma-Aldrich
Magnesium oxide, ≥99.99% trace metals basis
Sigma-Aldrich
Magnesium oxide, (single crystal substrate), ≥99.9% trace metals basis, <100>, L × W × thickness 10 mm × 10 mm × 0.5 mm
Sigma-Aldrich
Magnesium oxide, 99.99% trace metals basis
Sigma-Aldrich
Magnesium oxide, tested according to Ph. Eur., heavy
Sigma-Aldrich
Magnesium oxide, BioUltra, ≥97.0% (calcined substance, KT)
Sigma-Aldrich
Magnesium oxide, puriss., meets analytical specification of Ph. Eur., BP, USP, FCC, E 530, light, 98.0-100.5% (calc. for dried substance)
Sigma-Aldrich
Magnesium oxide, ≥99% trace metals basis, -325 mesh
Sigma-Aldrich
Magnesium oxide, light, 95%
Sigma-Aldrich
Magnesium oxide, -10-+50 mesh, 98%
Sigma-Aldrich
Magnesium oxide, ACS reagent, 97%