MilliporeSigma
  • The in vitro protective effects of curcumin and demethoxycurcumin in Curcuma longa extract on advanced glycation end products-induced mesangial cell apoptosis and oxidative stress.
The in vitro protective effects of curcumin and demethoxycurcumin in Curcuma longa extract on advanced glycation end products-induced mesangial cell apoptosis and oxidative stress.
Planta medica (2012-08-28)
Ji-ping Liu, Liang Feng, Mao-mao Zhu, Ru-Shang Wang, Ming-hua Zhang, Shao-ying Hu, Xiao-bin Jia, Jin-Jie Wu
ABSTRACT
Curcuma longa L. (CLL), a traditional herbal medicine, has been widely used for the prevention of diabetic vascular complications in recent years. However, the protective effects of curcuminoids in CLL on the AGEs-induced damage to mesangial cell are not fully understood. In this present study, dihydroethidium, superoxide dismutase kit, malondialdehyde kit, and acridine orange/ethidium bromide staining methods were used to evaluate the activities of curcumin and demethoxycurcumin (10(-11)-10(-9) M) on AGEs-induced oxidative stress and apoptosis, which were associated with the damage to mesangial cell. The results showed that these two compounds could significantly restore advanced glycation end products (AGEs)-induced apoptosis to normal levels (IC50 = 3.874 × 10(-11) M for curcumin and IC50 = 6.085 × 10(-11) M for demethoxycurcumin) and reduce remarkably reactive oxygen species generation in mesangial cell. Furthermore, curcumin and demethoxycurcumin dramatically elevated AGEs-decreased superoxide dismutase activity while significantly reducing AGEs-increased malondialdehyde content in cell culture supernatant. Our results suggest that both curcumin and demethoxycurcumin have a significant protective potential to the prevention of diabetic nephropathy.
Product Description
Supelco
Demethoxycurcumin, analytical standard
Sigma-Aldrich
Demethoxycurcumin, ≥98% (HPLC)
USP
Desmethoxycurcumin, United States Pharmacopeia (USP) Reference Standard