MilliporeSigma
  • Production of a calcium silicate cement material from alginate impression material.
Production of a calcium silicate cement material from alginate impression material.
Dental materials journal (2012-08-07)
Norimasa Washizawa, Hideaki Narusawa, Yukimichi Tamaki, Takashi Miyazaki
ABSTRACT
The purpose of this study was to synthesize biomaterials from daily dental waste. Since alginate impression material contains silica and calcium salts, we aimed to synthesize calcium silicate cement from alginate impression material. Gypsum-based investment material was also investigated as control. X-ray diffraction analyses revealed that although firing the set gypsum-based and modified investment materials at 1,200°C produced calcium silicates, firing the set alginate impression material did not. However, we succeeded when firing the set blend of pre-fired set alginate impression material and gypsum at 1,200°C. SEM observations of the powder revealed that the featured porous structures of diatomite as an alginate impression material component appeared useful for synthesizing calcium silicates. Experimentally fabricated calcium silicate powder was successfully mixed with phosphoric acid solution and set by depositing the brushite. Therefore, we conclude that the production of calcium silicate cement material is possible from waste alginate impression material.
Sigma-Aldrich
Celite® 503, CP
Sigma-Aldrich
Hyflo® Super-Cel®, CP
Supelco
Celite® 545 AW, reagent grade
Supelco
Celatom®, acid-washed, for use in Total Dietary Fiber Assay, TDF-100A
Sigma-Aldrich
Celite® 503, filter aid, treated with sodium carbonate, calcined
Sigma-Aldrich
Celpure® P65, meets USP/NF testing specifications
Sigma-Aldrich
Hyflo® Super Cel®, filter aid, flux calcined, treated with sodium carbonate
Sigma-Aldrich
Celite® 535, CP
Sigma-Aldrich
Diatomaceous earth non-washed, CP
Sigma-Aldrich
Calcium silicate, −200 mesh, 99%
Sigma-Aldrich
Celite® S, filter aid, dried, untreated
Sigma-Aldrich
Calcium silicate, purum, 12-22% Ca (as CaO) basis, ≥87% SiO2 basis
Sigma-Aldrich
Diatomaceous earth non-washed, powder
Sigma-Aldrich
Diatomaceous earth
Sigma-Aldrich
Filter agent, Celite® 545
Sigma-Aldrich
Celpure® P1000, meets USP/NF testing specifications
Sigma-Aldrich
Celite® 535, filter aid, flux calcined
Sigma-Aldrich
Celite® 545, filter aid, treated with sodium carbonate, flux calcined
Sigma-Aldrich
Celite® 545 AW, filter aid, acid washed, treated with sodium carbonate, flux calcined