MilliporeSigma
  • Base- and metal-free C-H direct arylations of naphthalene and other unbiased arenes with diaryliodonium salts.
Base- and metal-free C-H direct arylations of naphthalene and other unbiased arenes with diaryliodonium salts.
Chemical communications (Cambridge, England) (2012-08-03)
Susana Castro, Juan J Fernández, Rubén Vicente, Francisco J Fañanás, Félix Rodríguez
ABSTRACT
A novel metal-free direct arylation of unbiased arenes with diaryliodonium triflates in a process where the use of solvents and additives is not required has been established.
Sigma-Aldrich
Diphenyliodonium chloride, ≥98.0% (AT)
Sigma-Aldrich
Diphenyliodonium nitrate
Sigma-Aldrich
Diphenyliodonium hexafluorophosphate, ≥98%