MilliporeSigma
  • Characterizing the intrinsic bioremediation potential of 1,4-dioxane and trichloroethene using innovative environmental diagnostic tools.
Characterizing the intrinsic bioremediation potential of 1,4-dioxane and trichloroethene using innovative environmental diagnostic tools.
Journal of environmental monitoring : JEM (2012-07-25)
Sheau-Yun Dora Chiang, Rebecca Mora, William H Diguiseppi, Greg Davis, Kerry Sublette, Phillip Gedalanga, Shaily Mahendra
ABSTRACT
An intrinsic biodegradation study involving the design and implementation of innovative environmental diagnostic tools was conducted to evaluate whether monitored natural attenuation (MNA) could be considered as part of the remedial strategy to treat an aerobic aquifer contaminated with 1,4-dioxane and trichloroethene (TCE). In this study, advanced molecular biological and stable isotopic tools were applied to confirm in situ intrinsic biodegradation of 1,4-dioxane and TCE. Analyses of Bio-Trap® samplers and groundwater samples collected from monitoring wells verified the abundance of bacteria and enzymes capable of aerobically degrading TCE and 1,4-dioxane. Furthermore, phospholipid fatty acid analysis with stable isotope probes (PLFA-SIP) of the microbial community validated the ability for microbial degradation of TCE and 1,4-dioxane. Compound specific isotope analysis (CSIA) of groundwater samples for TCE resulted in δ(13)C values that indicated likely biodegradation of TCE in three of the four monitoring wells sampled. Results of the MNA evaluation showed that enzymes capable of aerobically degrading TCE and 1,4-dioxane were present, abundant, and active in the aquifer. Taken together, these results provide direct evidence of the occurrence of TCE and 1,4-dioxane biodegradation at the study site, supporting the selection of MNA as part of the final remedy at some point in the future.
Sigma-Aldrich
1,4-Dioxane solution, suitable for NMR (reference standard), 40% in benzene-d6 (99.6 atom % D), chromium(III) acetylacetonate 5 mg/mL, NMR tube size 3 mm × 8 in.
Sigma-Aldrich
1,4-Dioxane solution, suitable for NMR (reference standard), 40% in benzene-d6 (99.6 atom % D), NMR tube size 10 mm × 8 in.
Supelco
1,4-Dioxane solution, suitable for NMR (reference standard), 5 mM in benzene-d6 (99.6 atom % D), NMR tube size 3 mm × 8 in.
Sigma-Aldrich
1,4-Dioxane solution, suitable for NMR (reference standard), 10 mM in D2O ("100%", 99.96 atom % D), NMR tube size 5 mm × 7 in.
Supelco
1,4-Dioxane solution, suitable for NMR (reference standard), 5 mM in chloroform-d (99.8 atom % D), NMR tube size 3 mm × 8 in.
Sigma-Aldrich
1,4-Dioxane, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.5% (GC)
Sigma-Aldrich
1,4-Dioxane, suitable for HPLC, ≥99.5%
Sigma-Aldrich
1,4-Dioxane, ACS reagent, ≥99.0%, contains ≤25 ppm BHT as stabilizer
Sigma-Aldrich
1,4-Dioxane solution, suitable for NMR (reference standard), 40% in benzene-d6 (99.6 atom % D)
Supelco
1,4-Dioxane solution, suitable for NMR (reference standard), 40% in benzene-d6 (99.6 atom % D), NMR tube size 5 mm × 8 in.
Sigma-Aldrich
1,4-Dioxane solution, suitable for NMR (reference standard), 40% in benzene-d6 (99.6 atom % D), chromium(III) acetylacetonate 5 mg/mL, NMR tube size 5 mm × 8 in.
Supelco
1,4-Dioxane, analytical standard
Sigma-Aldrich
1,4-Dioxane, JIS special grade, ≥99.0%
Sigma-Aldrich
1,4-Dioxane, suitable for HPLC
Sigma-Aldrich
1,4-Dioxane, ACS reagent, ≥99.0%, contains <=25 ppm BHT as stabilizer
Sigma-Aldrich
1,4-Dioxane, ReagentPlus®, ≥99%, contains ≤25 ppm BHT as stabilizer
Sigma-Aldrich
1,4-Dioxane, anhydrous, 99.8%, contains <=25 ppm BHT as stabilizer
Sigma-Aldrich
1,4-Dioxane, SAJ first grade
Supelco
1,4-Dioxane solution, certified reference material, 2000 μg/mL in methanol, ampule of 1 mL