MilliporeSigma
  • Pharmacodynamics of ginsenosides: antioxidant activities, activation of Nrf2, and potential synergistic effects of combinations.
Pharmacodynamics of ginsenosides: antioxidant activities, activation of Nrf2, and potential synergistic effects of combinations.
Chemical research in toxicology (2012-07-12)
Constance Lay Lay Saw, Anne Yuqing Yang, David C Cheng, Sarandeep S-S Boyanapalli, Zheng-Yuan Su, Tin Oo Khor, Song Gao, Jingrong Wang, Zhi-Hong Jiang, Ah-Ng Tony Kong
ABSTRACT
Ginseng has long been used in Asian countries for more than 2000 years. Currently, in the "Western World or Western Medicines", many reports have indicated that they have used herbal medicines, and ginseng is one of the most popular herbs. Several recent reports have indicated that the antioxidant/antioxidative stress activities of ginseng play a role in the benefits of ginseng; however, the precise mechanism is lacking. The antioxidant response element (ARE) is a critical regulatory element for the expression of many antioxidant enzymes and phase II/III drug metabolizing/transporter genes, mediated by the transcription factor nuclear factor (erythroid-derived 2)-like 2 (Nrf2). The aim of this study was to examine the potential activation and synergism of Nrf2-ARE-mediated transcriptional activity between three common ginsenosides present in ginseng, ginsenoside Rb1 (Rb1), ginsenoside Rg1 (Rg1), and ginsenoside 20(S)-protopanaxatriol (20S). We tested whether these ginsenosides and their combinations could induce Nrf2-ARE activities in HepG2-C8 cells with stably transfected ARE luciferase reporter gene. Cell proliferation, antioxidant and ARE activities, Western blotting of Nrf2 protein, and qPCR of mRNA of Nrf2 were conducted for Rb1, Rg1, and 20S as well as the combinations of 20S with Rb1 or Rg1. To determine the combination effects, the combination index (CI) was calculated. Rb1 and Rg1 are relatively nontoxic to the cells, while 20S at 50 μM or above significantly inhibited the cell proliferation. Rb1, Rg1, or 20S induced total antioxidant activity and ARE activity in a concentration-dependent manner. Furthermore, combinations of 20S with either Rb1 or Rg1 induced total antioxidant and ARE activity synergistically. The induction of Nrf2 protein and mRNA was also found to be synergistic with the combination treatments. In summary, in this study, we show that ginsenosides Rb1, Rg1, and 20S possess antioxidant activity, transcriptionally activating ARE as well as the potential of synergistic activities. The Nrf2-ARE-mediated antioxidant pathway could play a role for the overall antioxidative stress activities, which could be important for ginseng's health beneficial effects such as cancer chemopreventive activities.
Product Description
Ginsenoside Rg1, primary reference standard
Supelco
Ginsenoside Rg1, analytical standard