MilliporeSigma
Polymeric material for anti-biofouling.
Colloids and surfaces. B, Biointerfaces (2012-07-07)
Chunfeng Ma, Hongjun Yang, Xi Zhou, Bo Wu, Guangzhao Zhang
ABSTRACT
Copolymerization of methyl methacrylate (MMA) and acrylate terminated poly(ethylene oxide-co-ethylene carbonate) (PEOC) macromonomer yields graft copolymer with PEOC side chains. By use of quartz crystal microbalance with dissipation (QCM-D), we have investigated the adsorption of proteins on the surface constructed by the copolymer in seawater. It shows the polymeric surface exhibits protein resistance depending on the content of the side chains. Marine field tests show the protein resistant surface can effectively inhibit marine biofouling for more than 12 weeks.
Sigma-Aldrich
Methyl methacrylate, SAJ first grade, ≥99.0%
Sigma-Aldrich
Methyl methacrylate, 99%, stabilized
Sigma-Aldrich
Methyl methacrylate, contains ≤30 ppm MEHQ as inhibitor, 99%
Supelco
Methyl methacrylate solution, certified reference material, 1000 μg/mL in methanol