MilliporeSigma
  • Effects of ethanol and dimethyl sulfoxide on the molecular organization of H2O as probed by 1-propanol.
Effects of ethanol and dimethyl sulfoxide on the molecular organization of H2O as probed by 1-propanol.
The journal of physical chemistry. B (2012-06-01)
Takeshi Morita, Peter Westh, Keiko Nishikawa, Yoshikata Koga
ABSTRACT
We characterized the effects of ethanol (ET) and dimethyl sulfoxide (DMSO) on H(2)O within a limited H(2)O-rich region by the 1-propanol (1P)-probing methodology developed by us earlier. The results are displayed on a two-dimensional map with twin coordinates: one pertaining to hydrophobicity and the other to hydrophilicity. The locus of ET on this map was at a point in between methanol (ME) and 2-propanol (2P) as expected from our earlier findings by thermodynamic studies. That for DMSO, however, was surprisingly more hydrophilic than ME. Similar to N-methyl groups discussed recently (J. Phys. Chem. B 2011, 115, 2995), it was argued that the methyl groups attached to the S atom are made susceptible for direct hydrogen bonding to the surrounding H(2)O molecules due possibly to the electronegativity of the S atom. In view of these findings, we suggest caution to be exercised for the conventional general trend of taking any methyl groups to be "hydrophobic."
Sigma-Aldrich
1-Propanol, JIS special grade
Sigma-Aldrich
1-Propanol, SAJ first grade, ≥99.0%
Sigma-Aldrich
1-Propanol, natural, ≥98%, FG
Supelco
1-Propanol, analytical standard
Sigma-Aldrich
1-Propanol, anhydrous, 99.7%
Sigma-Aldrich
1-Propanol, ≥99%, FG
Sigma-Aldrich
1-Propanol, suitable for HPLC, ≥99.9%
Sigma-Aldrich
1-Propanol, ACS reagent, ≥99.5%
Sigma-Aldrich
1-Propanol, ≥99% (GC), purum