MilliporeSigma
  • Gamma radiation measurements and Monte Carlo computations following myocardial perfusion imaging with 201Tl.
Gamma radiation measurements and Monte Carlo computations following myocardial perfusion imaging with 201Tl.
Radiation protection dosimetry (2012-05-23)
A Clouvas, S Xanthos
ABSTRACT
In the current study, the time-dependent retention of (201)Tl-thallous chloride (111 MBq) was measured in a 56-y-old man undergoing myocardial perfusion imaging. For 23 d following the (201)Tl injection, total-body retained activity was measured by (i) in situ gamma spectrometry using a portable high-purity germanium (HPGe) detector and (ii) ex vivo urine radioassay using a shielded HPGe detector. The time-dependent decrease in total-body activity followed a monoexponential function, exp(-0.011 t), with an excellent correlation (R(2)=0.9988) between the experimental data and the fitted values. The effective half-life, Teff, of (201)Tl (physical half-life, Tph: 72.9 h) was therefore 63 h and the biological half-life, Tb, 463 h=19.3 d, identical to those measured in the same patient in 1997 (i.e. 14 y ago). The time-dependent decrease in the urine activity concentration, which followed a monoexponential function, exp(-0.0115 t), corroborated the foregoing results. The correlation (R(2)=0.9939) between the experimental data and the fitted values was again excellent. The effective half-life, Teff, was 60.26 h and the biological half-life, Tb, 348 h=14.5 d. Monte Carlo simulation using a simple model of the patient as a unit-density cylinder filled with water and containing a uniform distribution of (201)Tl yielded photon flux results in reasonable agreement with the measured data.
Sigma-Aldrich
Thallium(I) chloride, 99.999% trace metals basis
Sigma-Aldrich
Thallium(I) chloride, 99%