MilliporeSigma
  • Plasma polymer films as an alternative to (PSS-PAH)n or (PSS-PDADMAC)n films to retain active enzymes in exponentially growing polyelectrolyte multilayers.
Plasma polymer films as an alternative to (PSS-PAH)n or (PSS-PDADMAC)n films to retain active enzymes in exponentially growing polyelectrolyte multilayers.
Colloids and surfaces. B, Biointerfaces (2012-05-23)
Cédric Amorosi, Marc Michel, Luc Avérous, Valérie Toniazzo, David Ruch, Vincent Ball
ABSTRACT
Compact and linearly growing polyelectrolyte multilayer films have been used to suppress desorption of drugs, nanoparticles or proteins from underlying polyelectrolyte multilayer films as well as to significantly change their mechanical properties. The polyelectrolyte based capping layers are however cumbersome to deposit and alternative barrier layers offering enzymatic retention in the films as well as permeability to small molecules, for example the substrates of the embedded enzymes, are highly desired. In this article we show that barrier layers made through atmospheric pressure dielectric barrier discharge polymerization of ethylene glycol dimethacrylate offer the opportunity to simultaneously suppress enzyme desorption from the underlying polyelectrolyte multilayer film and to ensure accessibility of the enzymatic substrate. This holds true when the barrier film totally covers the underlying film and as long is not too thick. When the plasma deposited barrier becomes 300 nm thick, the hydrolysis curve of the enzyme presents a lag phase typical of a diffusion-limited process.
Sigma-Aldrich
Poly(diallyldimethylammonium chloride) solution, average Mw <100,000 (very low molecular weight), 35 wt. % in H2O
Sigma-Aldrich
Poly(diallyldimethylammonium chloride) solution, average Mw 400,000-500,000 (high molecular weight), 20 wt. % in H2O
Sigma-Aldrich
Poly(diallyldimethylammonium chloride) solution, 20 wt. % in H2O
Sigma-Aldrich
Poly(diallyldimethylammonium chloride) solution, average Mw 200,000-350,000 (medium molecular weight), 20 wt. % in H2O