MilliporeSigma
  • Administration of estradiol, trenbolone acetate, and trenbolone acetate/estradiol implants alters adipogenic and myogenic gene expression in bovine skeletal muscle.
Administration of estradiol, trenbolone acetate, and trenbolone acetate/estradiol implants alters adipogenic and myogenic gene expression in bovine skeletal muscle.
Journal of animal science (2012-05-11)
K Y Chung, T J Baxa, S L Parr, L D Luqué, B J Johnson
ABSTRACT
Twenty crossbred yearling steers (421 kg) were used to evaluate the effects of implanting with trenbolone acetate (TBA; 120 mg), estradiol-17β (E(2); 25.7 mg), and a combination (120 mg of TBA and 24 mg of E(2)) on adipogenic and myogenic mRNA concentrations. Animals were blocked by BW and within each block were assigned to 1 of 4 treatments. Animals were housed and fed in individual pens with 5 animals per treatment. All animals were weighed weekly, and muscle biopsy samples were taken from the LM of each steer on d 0 (before implantation), 7, 14, and 28. Total RNA was isolated from each sample and real-time quantitative PCR was used to measure the quantity of C/EBPβ, PPARγ, stearoyl CoA desaturase (SCD), myogenin, and 3 isoforms of bovine myosin heavy chain (MHC) mRNA. Total BW gain from the 28-d period was adjusted to d 0 by use of covariant analysis, and steers in the implant groups tended (P = 0.09) to have increased BW gain compared with nonimplanted control steers. Analysis of the gene expression of MHC showed that neither implant nor day (P > 0.20) had a significant effect on the expression of type I or IIX MHC mRNA There was also no treatment effect (P > 0.20) on MHC-IIA and myogenin, but increasing days on feed increased (P = 0.05) the expression of MHC-IIA mRNA. Relative mRNA abundance of C/EBPβ, PPARγ, and SCD increased (P < 0.05) during days of feed but PPARγ decreased (P < 0.05) with the treatment of combined TBA/E(2) implant. Results of this study indicate that implanting with TBA, E(2), or both increased BW gain and decreased adipogenic gene expression of finishing steers without significantly affecting the concentration of type I, IIA, or IIX MHC mRNA. Increasing days on feed increased both MHC-IIA and adipogenic gene expression in bovine skeletal muscle biopsy samples. We conclude that administration of steroidal implants had no effect on the proportion of the 3 MHC mRNA isoforms but decreased C/EBPβ, PPARγ, and SCD mRNA in bovine skeletal muscle.
Product Description
Supelco
Trenbolone, analytical standard, for drug analysis
Supelco
Trenbolone solution, 1.0 mg/mL in acetonitrile, ampule of 1 mL, certified reference material, Cerilliant®