MilliporeSigma
Antiproliferative effect of oleuropein in prostate cell lines.
International journal of oncology (2012-04-10)
Rosaria Acquaviva, Claudia Di Giacomo, Valeria Sorrenti, Fabio Galvano, Rosa Santangelo, Venera Cardile, Silvia Gangia, Nicolantonio D'Orazio, Nader G Abraham, Luca Vanella
ABSTRACT
Currently, there is increasing interest in the in vivo protective effects of natural antioxidants found in dietary plants against oxidative damage caused by free radical species. Oxidative stress has been invoked as a causative agent in cancer and epidemiological data suggest that the consumption of fruits and vegetables may be associated with a lower incidence of cancer. The fruit of the Olea europaea L. and olive oil contain hundreds of phytochemicals and its extracts have recently been shown to exhibit antioxidant properties, due to the action of oleuropein. In view of these considerations, in this study, we investigated the effects of oleuropein on LNCaP and DU145 prostate cancer cell lines and on BPH-1 non-malignant cells. Oleuropein reduces cell viability and induces thiol group modifications, γ-glutamylcysteine synthetase, reactive oxygen species, pAkt and heme oxygenase-1. Exposing cell cultures to oleuropein induces an antioxidant effect on BPH-1 cells and a pro-oxidant effect on cancer cells. Our results confirm the beneficial properties of olive oil and oleuropein, suggesting its possible use as an adjuvant agent in the treatment of prostatitis, in order to prevent the transformation of hypertrophic to cancerous cells.
Sigma-Aldrich
Oleuropein, ≥80% (HPLC)
Supelco
Oleuropein, analytical standard
Sigma-Aldrich
Oleuropein, ≥98.0%