MilliporeSigma
  • Development of a highly sensitive method for the quantification of estrone and estradiol in serum by liquid chromatography tandem mass spectrometry without derivatization.
Development of a highly sensitive method for the quantification of estrone and estradiol in serum by liquid chromatography tandem mass spectrometry without derivatization.
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences (2012-03-21)
Tom Fiers, Bruno Casetta, Brigitte Bernaert, Eric Vandersypt, Martine Debock, Jean-Marc Kaufman
ABSTRACT
Measurement of estrone (E1) and estradiol (E2) values <1 pg/mL (3.7 pmol/L) is necessary for postmenopausal, pediatric and male serum samples. Until now this was rarely reached and only through derivatization which can present problems for estradiol. A very sensitive LC-MS/MS method was developed avoiding derivatization, convenient for large-scale studies. The desired sensitivity and specificity were achieved using ESI negative mode, LLE and a 2D chromatography consisting of a trapping column and a second dimension reverse-phase C8 analytical column. A mixture of an aqueous solution of ammonium fluoride at 0.2mM and methanol was used on the analytical column to further increase the sensitivity. Serum LOQ was <0.5 pg/mL (1.9 pmol/L) for E2 and E1 and recoveries ranged from 95 to 105%. No carry-over was detectable. Inter assay CV's were 4.0% at 21 pg/mL (77 pmol/L) for E2, 7.6% at 25 pg/mL (93 pmol/L) for E1. Comparison with commercial direct estrogen assays (Roche Diagnostics E170 for E2, Bioline RIA for E1) exposed analytical unsuitability (due to a combined lack of sensitivity and specificity) for the assay of male, postmenopausal or pediatric samples.
Sigma-Aldrich
Ammonium fluoride, ≥99.99% trace metals basis
Sigma-Aldrich
Ammonium fluoride, ACS reagent, ≥98.0%
Sigma-Aldrich
Ammonium hydrogen difluoride, SAJ special grade, ≥98.5%
Sigma-Aldrich
Ammonium hydrogen difluoride, 99.999% trace metals basis
Sigma-Aldrich
Ammonium hydrogen difluoride, reagent grade, 95%