MilliporeSigma
  • Doping of biogenic Pd catalysts with Au enables dechlorination of diclofenac at environmental conditions.
Doping of biogenic Pd catalysts with Au enables dechlorination of diclofenac at environmental conditions.
Water research (2012-03-13)
Simon De Corte, Tom Sabbe, Tom Hennebel, Lynn Vanhaecke, Bart De Gusseme, Willy Verstraete, Nico Boon
ABSTRACT
By using the metal reducing capacities of bacteria, Pd nanoparticles can be produced in a sustainable way ('bio-Pd'). These bio-Pd nanoparticles can be used as a catalyst in, for example, dehalogenation reactions. However, some halogenated compounds are not efficiently degraded using a bio-Pd catalyst. This study shows that the activity of bio-Pd can be improved by doping with Au(0) ('bio-Pd/Au'). In contrast with bio-Pd, bio-Pd/Au could perform the removal of the model pharmaceutical compound diclofenac from an aqueous medium in batch experiments at neutral pH and with H(2) as the hydrogen donor (first order decay constant of 0.078 ± 0.009 h(-1)). Dehalogenation was for both catalysts the only observed reaction. For bio-Pd/Au, a disproportional increase of catalytic activity was observed with increasing Pd-content of the catalyst. In contrast, when varying the Au-content of the catalyst, a Pd/Au mass ratio of 50/1 showed the highest catalytic activity (first order decay value of 0.52 ± 0.02 h(-1)). The removal of 6.40 μg L(-1) diclofenac from a wastewater treatment plant effluent using bio-Pd was not possible even after prolonged reaction time. However, by using the most active bio-Pd/Au catalyst, 43.8 ± 0.5% of the initially present diclofenac could be removed after 24 h. This study shows that doping of bio-Pd nanoparticles with Au(0) can be a promising approach for the reductive treatment of wastewaters containing halogenated contaminants.
Sigma-Aldrich
Diatrizoic acid, Iodine-containing contrast agent