MilliporeSigma
  • Suzuki coupling of potassium cyclopropyl- and alkoxymethyltrifluoroborates with benzyl chlorides.
Suzuki coupling of potassium cyclopropyl- and alkoxymethyltrifluoroborates with benzyl chlorides.
The Journal of organic chemistry (2012-03-07)
Virginie Colombel, Frederik Rombouts, Daniel Oehlrich, Gary A Molander
ABSTRACT
Efficient Csp(3)-Csp(3) Suzuki couplings have been developed with both potassium cyclopropyl- and alkoxymethyltrifluoroborates. Moderate to good yields have been achieved in the cross-coupling of potassium cyclopropyltrifluoroborate with benzyl chlorides possessing electron-donating or electron-withdrawing substituents. Benzyl chloride was also successfully cross-coupled to potassium alkoxymethyltrifluoroborates derived from primary, secondary, and tertiary alcohols.
Sigma-Aldrich
tert-Butanol, SAJ special grade, ≥99.0%
Sigma-Aldrich
tert-Butanol, SAJ first grade, ≥98.0%
Sigma-Aldrich
tert-Butanol, anhydrous, ≥99.5%
Sigma-Aldrich
Benzyl chloride, ReagentPlus®, 99%, contains ≤1% propylene oxide as stabilizer
Supelco
tert-Butanol, analytical standard
Sigma-Aldrich
tert-Butanol, puriss. p.a., ACS reagent, ≥99.7% (GC)
Sigma-Aldrich
tert-Butanol, TEBOL® 99, ≥99.3%
Sigma-Aldrich
tert-Butanol, ACS reagent, ≥99.0%
Sigma-Aldrich
tert-Butanol, ≥99% (GC)
Sigma-Aldrich
tert-Butanol, suitable for HPLC, ≥99.5%