MilliporeSigma
  • Multicenter phase II randomized study evaluating dose-response of antiperistaltic effect of L-menthol sprayed onto the gastric mucosa for upper gastrointestinal endoscopy.
Multicenter phase II randomized study evaluating dose-response of antiperistaltic effect of L-menthol sprayed onto the gastric mucosa for upper gastrointestinal endoscopy.
Digestive endoscopy : official journal of the Japan Gastroenterological Endoscopy Society (2012-02-22)
Naoki Hiki, Michio Kaminishi, Kenjiro Yasuda, Noriya Uedo, Masumi Kobari, Terufumi Sakai, Takashi Hiratsuka, Kyota Ohno, Hajime Honjo, Sachiyo Nomura, Naohisa Yahagi, Hisao Tajiri, Hiroaki Suzuki
ABSTRACT
Peppermint oil solution was found to be effective for reducing gastric spasm during upper gastrointestinal endoscopy. The aim of the present study was to assess whether the gastric peristalsis-suppressing effect is dose-dependently induced by L-menthol, the major constituent of peppermint oil, and to determine the recommended dose of an L-menthol preparation. In this phase II, multicenter, double-blind, dose-response study, 131 eligible patients were randomly assigned to receive 20 mL of 0.4% L-menthol (n = 32), 0.8% L-menthol (n = 35), 1.6% L-menthol (n = 30), or placebo (n = 34). The primary efficacy measure was the proportion of subjects with no peristalsis in two time periods, 75 to 105 s after treatment and immediately before the completion of endoscopy. The peristalsis-suppressing effect of L-menthol increased dose dependently (5.6%, 32.0%, 47.4% and 52.9% in the 0%, 0.4%, 0.8% and 1.6% groups, respectively: P < 0.001, one-tailed Cochran-Armitage trend test). As compared with the placebo group, the proportion of subjects with no peristalsis after administration was significantly higher in the 0.8% group (P = 0.015) and 1.6% group (P = 0.009). Adverse events in the L-menthol dose groups occurred with similar frequencies in the placebo group. L-menthol suppresses peristalsis in a dose-dependent manner, and the dose-response reaches a plateau at 0.8% L-menthol. Further Phase III studies are needed to establish the superiority of 0.8% L-menthol over placebo.
Sigma-Aldrich
Peppermint oil, natural, from Mentha piperita L.
Sigma-Aldrich
Peppermint oil, natural, FCC, FG
Sigma-Aldrich
Peppermint oil, purified by triple-distillation, FCC, FG