MilliporeSigma
Regulation of xylanase biosynthesis in Bacillus cereus BSA1.
Applied biochemistry and biotechnology (2012-01-10)
Asish Mandal, Sanjay Kar, Pradeep Kumar Das Mohapatra, Chiranjit Maity, Bikas Ranjan Pati, Keshab Chandra Mondal
ABSTRACT
Microbial xylanases have a promising biotechnological potential to be used in industries. In this study, regulation of xylanase production was examined in Bacillus cereus BSA1. Xylanase production was induced by xylan. The enzyme production further increased in the presence of xylose and arabinose in very low concentration with addition of xylan (0.5% up to 6.02 U/ml). Addition of glucose (about 0.1%) to the media supplemented with xylan repressed xylanase production. Even higher concentration (>0.1%) of xylose and arabinose repressed xylanase biosynthesis. Glucose-mediated repression was partially relived by addition of cyclic adenosine monophosphate. Chemical like 2-4-dinitrophenol, which can inhibit adenosine triphosphate synthesis in cell, repressed xylanase synthesis and it suggested xylanase synthesis to be an energy dependent process.
Product Description
Supelco
2,4-Dinitrophenol solution, reference material, 5000 μg/mL in methanol
Sigma-Aldrich
2,4-Dinitrophenol, SAJ special grade, ≥98.0%
Sigma-Aldrich
2,4-Dinitrophenol, moistened with water, ≥98.0%
Supelco
2,4-Dinitrophenol, PESTANAL®, analytical standard