MilliporeSigma
  • Development and characterization of zein-based microcarrier system for sustained delivery of aceclofenac sodium.
Development and characterization of zein-based microcarrier system for sustained delivery of aceclofenac sodium.
AAPS PharmSciTech (2011-12-15)
K Karthikeyan, Rachita Lakra, Rama Rajaram, Purna Sai Korrapati
ABSTRACT
Nonsteroidal anti-inflammatory drugs (NSAIDs) induce gastric injury on long-term usage. This study aims at reducing the side effect of NSAIDs by encapsulating in zein, an acid-resistant biopolymer. Aceclofenac-loaded zein microspheres were prepared by emulsification and solvent evaporation method. The stability of zein microspheres at gastric pH retarded the release of the entrapped drug and hence reduces the possibility of gastric injury. However, the in vitro release of aceclofenac was sustained up to 72 h at intestinal pH. Thus, zein microspheres pave the way for the development of safe and sustained delivery system for NSAIDs thereby achieving the desired therapeutic potential with reduced side effects for chronic inflammatory disorders.
Sigma-Aldrich
Aceclofenac, ≥98% (HPLC)
Aceclofenac for peak identification, European Pharmacopoeia (EP) Reference Standard