MilliporeSigma
  • The effects of oxaloacetate on hydrogen peroxide generation from ascorbate and epigallocatechin gallate in cell culture media: potential for altering cell metabolism.
The effects of oxaloacetate on hydrogen peroxide generation from ascorbate and epigallocatechin gallate in cell culture media: potential for altering cell metabolism.
Biochemical and biophysical research communications (2011-12-15)
Lee Hua Long, Barry Halliwell
ABSTRACT
Several phenolic compounds as well as ascorbate can oxidise in certain cell culture media (especially Dulbecco's modified Eagle's medium (DMEM)) to generate hydrogen peroxide. Addition of oxaloacetate decreased the levels of H(2)O(2) detected and the oxaloacetate was depleted. Oxaloacetate was approximately as effective as pyruvate in decreasing H(2)O(2) levels and more effective than α-ketoglutarate. Our data raise important issues to consider when interpreting the behaviour and metabolism of cells in culture (which are both altered by the oxidative stress of cell culture) and their apparent response to addition of autooxidisable compounds such as ascorbate and epigallocatechin gallate.
Sigma-Aldrich
Oxaloacetic acid, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥97% (HPLC)
Sigma-Aldrich
Oxaloacetic acid, powder, suitable for hybridoma
Sigma-Aldrich
Oxaloacetic acid, ≥97% (HPLC)