MilliporeSigma
  • Evaluation of the developmental toxicity of leucomalachite green administered orally to rats.
Evaluation of the developmental toxicity of leucomalachite green administered orally to rats.
Food and chemical toxicology : an international journal published for the British Industrial Biological Research Association (2011-10-19)
Hongping Wan, Shaoping Weng, Lizhen Liang, Qihua Lu, Jianguo He
ABSTRACT
Malachite green (MG) had been extensively used worldwide in the past few decades as an effective fungicide, bactericide, ectoparasiticide, and antiprotozoan on fish. It is rapidly and extensively metabolized to leucomalachite green (LMG) in fishes. To study the developmental toxicity of LMG, we evaluated the maternal and embryo/fetal effects of LMG orally administered to Sprague-Dawley rats once daily from day 6-15 of gestation at dose levels of 0, 10, 80, and 160 mg/kg/day. In the current investigation, the unmetabolized compound (LMG) was detected in the plasma of the dams treated with LMG and in the amniotic fluid. At dose levels of 80 and 160 mg/kg/day, LMG induced maternal toxicity, which consisted of severe reduction in maternal weight and food consumption during the treatment period. Doses of 80 and 160 mg/kg/day induced fetal skeletal abnormalities. The major skeletal defects were bipartite ossification of the thoracic centrum. LMG at 160 mg/kg/day also induced marked embryolethality and visceral abnormalities. Based on the results of this study, a dose level of 10mg/kg/day is considered the no-observed-adverse-effect-level (NOAEL) for maternal and fetal developmental toxicity in rats.
Product Description
Supelco
Leucomalachite Green, analytical standard
Sigma-Aldrich
Leucomalachite Green, powder