MilliporeSigma
  • Spectrometric study on the interaction of dodecyltrimethylammonium bromide with curcumin.
Spectrometric study on the interaction of dodecyltrimethylammonium bromide with curcumin.
Langmuir : the ACS journal of surfaces and colloids (2011-10-19)
Dan Ke, Xiaoyong Wang, Qianqian Yang, Yumeng Niu, Shaohu Chai, Zhiyun Chen, Xueqin An, Weiguo Shen
ABSTRACT
The interaction between dodecyltrimethylammonium bromide (DTAB) and curcumin has been studied in pH 5.0 sodium phosphate buffer using absorption and fluorescence measurements. With increasing DTAB concentration (C(DTAB)) from 0 to 20 mM, the absorption peak of curcumin at 430 nm, corresponding to the conjugated structure of curcumin, first weakens gradually into a shoulder but increases back into one peak with much higher absorption intensity. On the contrary, as C(DTAB) increases, the initial small absorption shoulder of curcumin at 355 nm, corresponding to the feruloyl unit of curcumin, first increases gradually into a clear peak but decreases back into one shoulder until almost disappeared finally. By remaining at nearly the same wavelength, the fluorescence of curcumin first decreases at C(DTAB) lower than 5 mM and then increases gradually up to C(DTAB) = 10 mM, which is followed by sharp increases of fluorescence intensity with marked blue-shifts at higher C(DTAB). The values of anisotropy and microviscosity of curcumin obtained from the fluorescence polarization technique also showed pronounced changes at different surfactant concentrations. The interaction mechanisms of DTAB with curcumin have been presented at low, intermediate, and high surfactant concentrations, which is relating to interaction forces, surfactant aggregations, as well as structural alterations of curcumin.
Sigma-Aldrich
Dodecyltrimethylammonium bromide, BioXtra, ~99%
Sigma-Aldrich
Dodecyltrimethylammonium bromide, ≥98%
Supelco
Dodecyltrimethylammonium bromide, suitable for ion pair chromatography, LiChropur, ≥98.5% (AT)
Sigma-Aldrich
Dodecyltrimethylammonium chloride, purum, ≥98.0% anhydrous basis (AT)
Sigma-Aldrich
Dodecyltrimethylammonium chloride, ≥99.0% (AT)