MilliporeSigma
  • Chemical enhancement of footwear impressions in blood on fabric - part 2: peroxidase reagents.
Chemical enhancement of footwear impressions in blood on fabric - part 2: peroxidase reagents.
Science & justice : journal of the Forensic Science Society (2011-09-06)
Kevin J Farrugia, Kathleen A Savage, Helen Bandey, Tomasz Ciuksza, Niamh Nic Daéid
ABSTRACT
This study investigates the optimisation of peroxidase based enhancement techniques for footwear impressions made in blood on various fabric surfaces. Four different haem reagents: leuco crystal violet (LCV), leuco malachite green (LMG), fluorescein and luminol were used to enhance the blood contaminated impressions. The enhancement techniques in this study were used successfully to enhance the impressions in blood on light coloured surfaces, however, only fluorescent and/or chemiluminescent techniques allowed visualisation on dark coloured fabrics, denim and leather. Luminol was the only technique to enhance footwear impressions made in blood on all the fabrics investigated in this study.
Product Description
Supelco
Leucomalachite Green, analytical standard
Sigma-Aldrich
Leucomalachite Green, powder