MilliporeSigma
  • Combined chemical and topographical guidance cues for directing cytoarchitectural polarization in primary neurons.
Combined chemical and topographical guidance cues for directing cytoarchitectural polarization in primary neurons.
Biomaterials (2011-09-03)
Adrienne C Greene, Cody M Washburn, George D Bachand, Conrad D James
ABSTRACT
Chemical and topographical cues can be used to guide dissociated neurons into user-defined network geometries on artificial substrates, yet control of neuron polarity (differentiation into axons and dendrites) remains an elusive goal. We developed a dual guidance cue strategy for directing morphological maturity in neurons in vitro using combined chemical and topographical guidance cues on glass substrates. The surface chemistry provides chemical attraction and repulsion for controlling neuron placement and outgrowth, while the topography provides additional surface area for neuron attachment. Poly-l-lysine (PLL) was adsorbed into etched trenches in glass substrates, and an acetone liftoff process was used to produce bifunctional surfaces with a hydrophobic hexamethyldisilazane (HMDS) background and trench patterns of PLL. We examined the cytoarchitectural polarization of dissociated hippocampal pyramidal neurons on guidance cues designed to promote rapid outgrowth of neurites onto continuous line features and delayed neurite outgrowth onto interrupted line features. An optimum distance of approximately 5 μm between the cell body attachment node and the first interrupted line guidance cue led to specific cytoarchitectural polarization of ≥60% of neurons by 3 days of culture in vitro.
Sigma-Aldrich
Lithium bis(trimethylsilyl)amide, 97%
Sigma-Aldrich
Tris[N,N-bis(trimethylsilyl)amide]yttrium
Sigma-Aldrich
Tris[N,N-bis(trimethylsilyl)amide]samarium(III), 98%
Sigma-Aldrich
Potassium bis(trimethylsilyl)amide solution, 1 M in THF
Sigma-Aldrich
Potassium bis(trimethylsilyl)amide solution, 1.0 M in 2-methyltetrahydrofuran
Sigma-Aldrich
Lithium bis(trimethylsilyl)amide solution, 1.5 M in THF
Sigma-Aldrich
Lithium bis(trimethylsilyl)amide solution, 1.0 M in THF
Sigma-Aldrich
Sodium bis(trimethylsilyl)amide solution, 40% in THF
Supelco
Hexamethyldisilazane, for GC derivatization, LiChropur, ≥99.0% (GC)
Sigma-Aldrich
Hexamethyldisilazane, produced by Wacker Chemie AG, Burghausen, Germany, ≥97.0% (GC)
Sigma-Aldrich
Potassium bis(trimethylsilyl)amide, 95%
Sigma-Aldrich
Hexamethyldisilazane, reagent grade, ≥99%
Sigma-Aldrich
Lithium bis(trimethylsilyl)amide solution, 1.0 M in hexanes
Sigma-Aldrich
Potassium bis(trimethylsilyl)amide solution, 0.5 M in toluene
Sigma-Aldrich
Sodium bis(trimethylsilyl)amide solution, 0.6 M in toluene
Sigma-Aldrich
Lithium bis(trimethylsilyl)amide solution, 1 M in tert-butyl methyl ether
Sigma-Aldrich
Sodium bis(trimethylsilyl)amide, 95%
Sigma-Aldrich
Sodium bis(trimethylsilyl)amide solution, 1.0 M in THF
Sigma-Aldrich
Lithium bis(trimethylsilyl)amide solution, 0.5 M in 2-methyltetrahydrofuran
Sigma-Aldrich
Lithium bis(trimethylsilyl)amide solution, 1 M in toluene
Sigma-Aldrich
Hexamethyldisilazane, ReagentPlus®, 99.9%