MilliporeSigma
  • Analyzing velocity map images to distinguish the primary methyl photofragments from those produced upon C-Cl bond photofission in chloroacetone at 193 nm.
Analyzing velocity map images to distinguish the primary methyl photofragments from those produced upon C-Cl bond photofission in chloroacetone at 193 nm.
The Journal of chemical physics (2011-07-27)
Bridget W Alligood, Daniel B Straus, Laurie J Butler
ABSTRACT
We use a combination of crossed laser-molecular beam scattering experiments and velocity map imaging experiments to investigate the three primary photodissociation channels of chloroacetone at 193 nm: C-Cl bond photofission yielding CH(3)C(O)CH(2) radicals, C-C bond photofission yielding CH(3)CO and CH(2)Cl products, and C-CH(3) bond photofission resulting in CH(3) and C(O)CH(2)Cl products. Improved analysis of data previously reported by our group quantitatively identifies the contribution of this latter photodissociation channel. We introduce a forward convolution procedure to identify the portion of the signal, derived from the methyl image, which results from a two-step process in which C-Cl bond photofission is followed by the dissociation of the vibrationally excited CH(3)C(O)CH(2) radicals to CH(3) + COCH(2). Subtracting this from the total methyl signal identifies the methyl photofragments that result from the CH(3) + C(O)CH(2)Cl photofission channel. We find that about 89% of the chloroacetone molecules undergo C-Cl bond photofission to yield CH(3)C(O)CH(2) and Cl products; approximately 8% result in C-C bond photofission to yield CH(3)CO and CH(2)Cl products, and the remaining 2.6% undergo C-CH(3) bond photofission to yield CH(3) and C(O)CH(2)Cl products.
Sigma-Aldrich
Chloroacetone, 95%
Sigma-Aldrich
Chloroacetone, Wacker Chemie AG, ≥96.0% (GC)