MilliporeSigma
Lithium-induced sinus node dysfunction at therapeutic serum levels.
The National medical journal of India (2011-07-27)
L K Singh, S K Praharaj, S K Munda, C R J Khess
ABSTRACT
Lithium-induced cardiotoxicity, though rare at therapeutic levels, has been reported frequently in overdoses. We report a patient who developed sinus bradycardia while being treated with lithium carbonate even though the serum lithium levels were within the therapeutic range. It reversed following withdrawal of lithium and did not reappear with subsequent treatment with valproate.
Sigma-Aldrich
Lithium sulfate, purum p.a., ≥98.0% (T)
Sigma-Aldrich
Lithium sulfate, ≥98.5% (titration)
Sigma-Aldrich
Lithium sulfate, ≥99.99% trace metals basis