MilliporeSigma
  • Adverse effects of chromium oxide nano-particles on seed germination and growth in Triticum aestivum L.
Adverse effects of chromium oxide nano-particles on seed germination and growth in Triticum aestivum L.
Journal of biomedical nanotechnology (2011-04-14)
Poornima Vajpayee, Imrana Khatoon, Chandra Bali Patel, Gulshan Singh, Kailash Chand Gupta, Rishi Shanker
ABSTRACT
In this study, seeds of Triticum aestivum L. (Poaceae) were exposed to 0-100 microg/mL chromium oxide nanoparticles (Cr2O3, Nps) to study the phytotoxic effects on seed germination and seedling growth. It has been observed that 25-100 microg/mL Cr2O3, Nps inhibited the seed germination and seedling growth in concentration dependent manner. The present study suggests that release of Cr2O3, Nps in environment may adversely affect the wheat production.
Sigma-Aldrich
Chromium(III) oxide, SAJ first grade, ≥99.0%
Sigma-Aldrich
Chromium(III) oxide, nanopowder, <100 nm particle size (TEM), 98% trace metals basis
Sigma-Aldrich
Chromium(III) oxide, powder, 99.9% trace metals basis
Sigma-Aldrich
Chromium(III) oxide, powder, ≥98%