MilliporeSigma
  • Distribution and accumulation of endocrine-disrupting chemicals and pharmaceuticals in wastewater irrigated soils in Hebei, China.
Distribution and accumulation of endocrine-disrupting chemicals and pharmaceuticals in wastewater irrigated soils in Hebei, China.
Environmental pollution (Barking, Essex : 1987) (2011-04-12)
Feng Chen, Guang-Guo Ying, Ling-Xiao Kong, Li Wang, Jian-Liang Zhao, Li-Jun Zhou, Li-Juan Zhang
ABSTRACT
This study investigated the occurrence of 43 emerging contaminants including 9 endocrine-disrupting chemicals and 34 pharmaceuticals in three sites in Hebei Province, north China. Each site has a wastewater irrigated plot and a separate groundwater irrigated plot for comparison purpose. The results showed that the concentrations of the target compounds in the wastewater irrigated soils were in most cases higher than those in the groundwater irrigated soils. Among the 43 target compounds, nine compounds bisphenol-A, triclocarban, triclosan, 4-nonylphenol, salicylic acid, oxytetracycline, tetracycline, trimethoprim and primidone were detected at least once in the soils. Preliminary environmental risk assessment showed that triclocarban might pose high risks to terrestrial organisms while the other detected compounds posed minimal risks. Irrigation with wastewater could lead to presence or accumulation of some emerging contaminants to some extent in irrigated soils.
Product Description
Supelco
Primidone, analytical standard
Supelco
Primidone solution, 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®