MilliporeSigma
  • The sequence selectivity of DNA-targeted 9-aminoacridine cisplatin analogues in a telomere-containing DNA sequence.
The sequence selectivity of DNA-targeted 9-aminoacridine cisplatin analogues in a telomere-containing DNA sequence.
Journal of biological inorganic chemistry : JBIC : a publication of the Society of Biological Inorganic Chemistry (2011-04-06)
Moumita Paul, Vincent Murray
ABSTRACT
In this study, the detailed DNA sequence specificity of four acridine Pt complexes was examined and compared with that of cisplatin. The DNA sequence specificity was determined in a telomere-containing DNA sequence using a polymerase stop assay, with a fluorescent primer and an automated capillary DNA sequencer. The Pt compounds included an acridine intercalating moiety that was modified to give a 9-aminoacridine derivative, a 7-methoxy-9-aminoacridine derivative, a 7-fluoro-9-aminoacridine derivative and a 9-ethanolamine-acridine derivative. Compared with cisplatin, the DNA sequence specificity was most altered for the 7-methoxy-9-aminoacridine compound, followed by the 9-aminoacridine derivative, the 7-fluoro-9-aminoacridine compound and the 9-ethanolamine-acridine derivative. The DNA sequence selectivity for the four acridine Pt complexes was shifted away from runs of consecutive guanines towards single guanine bases, especially 5'-GA dinucleotides and sequences that contained 5'-CG. The sequence specificity was examined in telomeric and non-telomeric DNA sequences. Although it was found that telomeric DNA sequences were extensively damaged by the four acridine Pt complexes, there was no extra preference for telomeric sequences.
Product Description
Supelco
9-Aminoacridine, ≥99.5% (HPLC)