MilliporeSigma
Controlling biofilm with evidence-based dentifrices.
Compendium of continuing education in dentistry (Jamesburg, N.J. : 1995) (2011-04-06)
Sebastian G Ciancio
ABSTRACT
This review summarizes research that has assessed the effectiveness of various antimicrobial-containing dentifrices in preventing and/or reducing a number of oral health problems facing our patients today. The results of these studies indicate that, when compared with a conventional fluoride dentifrice, the triclosan/copolymer/fluoride dentifrice is the one with the most evidence to support its ability to deliver significant oral health benefits with no adverse effects. The benefits maybe summarized as follows: improved levels ofsupragingival plaque control; improved gingival health; reducedlikelihood of gingivitis progressing to periodontitis; arrest progression of periodontitis; prevention of supragingival calculus; and reduction in oral malodor. With increased interest in the association of oral health with systemic health, this dentifrice is well-positioned to help reduce the likelihood of gingivitis establishing itself and possibly developing into periodontitis (Figure 1). It also has the potential to have beneficial effects on general health because of its anti-inflammatory properties. Based on the results presented in this article, it is clear that the general population can derive significant clinical benefits from the daily use of a triclosan/copolymer/fluoride dentifrice. The dental profession should feel confident to recommend its use to patients to improve oral health and maintain or promote overall health.
Sigma-Aldrich
Poly(methyl vinyl ether-alt-maleic anhydride), average Mw ~216,000, average Mn ~80,000
Sigma-Aldrich
Poly(methyl vinyl ether-alt-maleic anhydride), average Mw ~1,080,000, average Mn ~311,000