MilliporeSigma
  • HPLC analysis of kaempherol and quercetin derivatives isolated by different extraction techniques from plant matrix.
HPLC analysis of kaempherol and quercetin derivatives isolated by different extraction techniques from plant matrix.
Journal of AOAC International (2011-03-12)
Krystyna Skalicka-Woźniak, Janusz Szypowski, Kazimierz Głowniak
ABSTRACT
Soxhlet extraction, ultrasound extraction, and accelerated solvent extraction (ASE), followed by RP-HPLC with a photodiode array detector was used for the determination of flavonoids in fruits of Peucedanum alsaticum. Three compounds were identified: a kaempherol derivative (astragalin), and two quercetin derivatives (quercitrin and hiperoside). The highest extraction yields of the selected compounds were obtained by use of exhaustive ASE.
Product Description
Quercitrin, primary reference standard
Sigma-Aldrich
Kaempferol 3-β-D-glucopyranoside, ≥97.0% (HPLC)