MilliporeSigma
  • Signal pathway of hippocampal apoptosis and cognitive impairment of mice caused by cerium chloride.
Signal pathway of hippocampal apoptosis and cognitive impairment of mice caused by cerium chloride.
Environmental toxicology (2011-03-09)
Zhe Cheng, Na Li, Jie Cheng, Renping Hu, Guodong Gao, Yaling Cui, Xiaolan Gong, Ling Wang, Fashui Hong
ABSTRACT
Experimental studies have demonstrated that lanthanides could impair cognitive functions of children and animals, but very little is known about the hippocampal apoptosis and its molecular mechanism. The study investigated the signal pathway of hippocampal apoptosis induced by intragastric administration of CeCl(3) for 60 consecutive days. It showed that cerium had been significantly accumulated in the mouse hippocampus, and CeCl(3) caused hippocampal apoptosis and impairment of spatial recognition memory of mice. CeCl(3) effectively activated caspase-3 and -9, inhibited Bcl-2, and increased the levels of Bax and cytochrome c, promoted accumulation of reactive oxygen species in the mouse hippocampus. It implied that CeCl(3)-induced apoptosis in the mouse hippocampus could be triggered via mitochondrion-mediated pathway. Our findings suggest the need for great caution to handle the lanthanides for workers and consumers.
Sigma-Aldrich
Cerium(III) chloride heptahydrate, 99.9% trace metals basis
Sigma-Aldrich
Cerium(III) chloride heptahydrate, purum p.a., ≥98.0% (AT)
Sigma-Aldrich
Cerium(III) chloride heptahydrate, 99.999% trace metals basis
Sigma-Aldrich
Cerium(III) chloride, AnhydroBeads, −10 mesh, 99.9%
Sigma-Aldrich
Cerium(III) chloride, AnhydroBeads, −10 mesh, ≥99.99% trace metals basis